
2,5-Difluorobenzenesulphonamide ≥98.0% (by GC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:120022-63-1
:
Formula:C₆H₅F₂NO₂S
MDL Number:MFCD00798115
:193.17 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:135.0 to 139.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together