
5-Bromo-1H-indazole ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:53857-57-1
:
Formula:C₇H₅BrN₂
MDL Number:MFCD00839493
:197.03 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:130.0 to 134.0 deg-C
Specification Test Results
Frequently Bought Together