
6-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24549-06-2
:
Formula:C₉H₁₃N
MDL Number:MFCD00007752
:135.21
Boiling Pt:231 °C (1013 hPa)
Melting Pt:–33 °C
Density:0,969 g/cm³ (20 °C)
Flash Pt:111°C
EINECS:246-309-6
:GHS07
Specification Test Results
- Appearance:Colorless to Light red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together