Ammonium peroxodisulphate (APS), powder for electrophoresis
RC-017
:
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
:
:228.2 g/mol
:13,00545
:120 °C
:1.98 g/cm³ (20 °C)
:Ambient
:231-786-5
:1444
:5.1,III
:GHS03
:GHS08
:GHS07
:7727-54-0
:(NH₄)₂S₂O₈
:MFCD00003390