HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid)
RC-121
:
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
- Environmentally Preferable:
: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
:
:13,04672
:210…215 °C
:1.448 g/cm³ (20 °C)
:238.31 g/mol
:Ambient
:230-907-9
:7365-45-9
:C₈H₁₈N₂O₄S
:MFCD00006158