L(+)-Diethyl tartrate ≥98.0% (by GC)
: TCI
:87-91-2
:
:C₈H₁₄O₆
:MFCD00009143
:206.2 g/mol
:14,03855
:162°C
:18.7 °C
:1,21 g/cm³ (20 °C)
:152°C
:201-783-3
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Specific rotation [a]20/D:+7.6 to +7.9 deg (neat)
Frequently Bought Together