Order Entry
Italy
ContactUsLinkComponent
 
 
4-(2-Pyridyl)benzaldehyde ≥95.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
4-(2-Pyridyl)benzaldehyde ≥95.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %
  • Melting point:
    48.0 to 53.0 deg-C

 

Frequently Bought Together