
2-(p-Tolyl)pyridine ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4467-06-5
:
Formula:C₁₂H₁₁N
MDL Number:MFCD00006281
:169.22 g/mol
Boiling Pt:157°C
EINECS:224-734-8
:GHS07
Specification Test Results
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together