
(2-Carboxyethyl)triphenylphosphonium bromide 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:51114-94-4
:
Formula:C₂₁H₂₀BrO₂P
MDL Number:MFCD00031698
:415.2601 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Powder and/or chunks
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together