



Methylamine 2 M in THF
Catalog # H27664.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:31.06 g/mol
Merck Index:14,6014
Density:0.861
Flash Pt:<–34 °C (–29 °F)
Storage Temperature:Ambient
EINECS:200-820-0
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS08
:GHS07
CAS Number:74-89-5
Formula:CH₅N
MDL Number:MFCD00008104