



Trimetilamina 1 M in THF
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-50-3
:
Formula:C₃H₉N
MDL Number:MFCD00008327
Density:0,850
Storage Temperature:Ambiente
EINECS:200-875-0
UN:2924
ADR:3,II
:59.11 g/mol
:GHS02
:GHS05
:GHS08
:GHS07
Related Information
Frequently Bought Together