



Trimethylamine ∼13% (w/w) ∼2 mol/L in THF
: TCI
:75-50-3
:
:C₃H₉N
:MFCD00008327
:14,09710
:3,II
:59.11 g/mol
:200-875-0
:2924
:GHS02
:GHS05
:GHS07
:GHS08
:Refrigerator
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):13.0 to 15.5 w/w%
Frequently Bought Together