1,3-Di(p-tolyl)-2-thiourea ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:621-01-2
:
Formula:C₁₅H₁₆N₂S
MDL Number:MFCD00008541
EINECS:210-665-0
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Melting point:177.0 to 181.0 deg-C
Specification Test Results
Frequently Bought Together