
(3-Phenylpropyl)triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7484-37-9
:
Formula:C₂₇H₂₆BrP
MDL Number:MFCD00051886
:461.39 g/mol
Melting Pt:208…212 °C
EINECS:231-289-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together