
Ethyl pentafluorobenzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4522-93-4
:
Formula:C₉H₅F₅O₂
MDL Number:MFCD00039211
:240.13 g/mol
Boiling Pt:112…114 °C
Density:1.431
Flash Pt:80 °C (176 °F)
EINECS:224-854-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together