Order Entry
Italy
ContactUsLinkComponent
D-(+)-Cellobiose octaacetate ≥98.0% (by GC)
D-(+)-Cellobiose octaacetate ≥98.0% (by GC)
Catalog # TCIAC0861-25G
Supplier:  TCI
CAS Number:  
undefined
D-(+)-Cellobiose octaacetate ≥98.0% (by GC)
Catalog # TCIAC0861-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    230.0 to 234.0 deg-C
  • Specific rotation [a]20/D:
    +38.0 to +42.0 deg (C=6, CHCl3)
  • Solubility in Chloroform:
    almost transparency

Specification Test Results

About this item

 :
 :678.59 g/mol
Storage Temperature:Ambient
EINECS:226-304-5
CAS Number:5346-90-7
Formula:C₂₈H₃₈O₁₉
MDL Number:MFCD00009600