Order Entry
Italy
ContactUsLinkComponent
2,4'-Dihydroxybenzophenone ≥98.0% (by GC)
Irritant
2,4'-Dihydroxybenzophenone ≥98.0% (by GC)
Catalog # TCIAD2664-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,4'-Dihydroxybenzophenone ≥98.0% (by GC)
Catalog # TCIAD2664-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    146.0 to 150.0 deg-C

Specification Test Results

About this item

 :
 :GHS07
CAS Number:606-12-2
Formula:C₁₃H₁₀O₃
MDL Number:MFCD02181674