4,4'-Dichlorobenzophenone-[D8] 100 µg/ml in acetone, Dr Ehrenstorfer
EHERXA12410100AC
:
- Pack type:Glass bottle
- Pk:1,1 mL
- CAS No.:1219806-01-5
- Description:4,4'-Dichlorobenzophenone-[D8] 100 µg/ml in acetone
- Formula:C₁₃₂H₈Cl₂O
- Description:4,4'-Dichlorobenzophenone D8