Order Entry
Italy
ContactUsLinkComponent
1,1-Dimethyl-4-phenylpiperazinium iodide ≥98.0% (by HPLC)
Toxic
1,1-Dimethyl-4-phenylpiperazinium iodide ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Toxic
1,1-Dimethyl-4-phenylpiperazinium iodide ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

Frequently Bought Together