Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Trypan blue

Glass bottle, 25 g
Catalog # TCIAT0556-25G
Health
Supplier:   TCI
Trypan blue
Catalog # TCIAT0556-25G
Supplier:  TCI
CAS Number:  72-57-1
Trypan blue
Trypan blue

Glass bottle, 25 g

CAS Number:  72-57-1
Price
29,00 €
Log in for availability
1 * 25 g
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)
  • CAS Number:72-57-1
  • Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
  • MDL Number:MFCD00003969
  • Molecular Weight:960.82 g/mol
  • Merck Index:14,09792
  • EINECS:200-786-7
Specifications

Specification Test Results

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Lambda max.:
    594.0 to 602.0 nm (H2O)
  • Absorbance (E1%1cm):
    min. 600 (H2O, 594.0 to 602.0 nm)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.