Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trypan Blue
Trypan blue
Trypan blue
Trypan blue
KOEHCN76.2
:
KOEHLER TECHNISCHE PRODUKTEN
:
Trypan blue
KOEHCN76.2
:
KOEHLER TECHNISCHE PRODUKTEN
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
960.82 g/mol
:
200-786-7
:
72-57-1
:
C₃₄H₂₄N₆Na₄O₁₄S₄
:
MFCD00003969