Order Entry
Italy
ContactUsLinkComponent
Trypan blue for microscopy, Sigma-Aldrich®
Health
Trypan blue for microscopy, Sigma-Aldrich®
Catalog # 1.11732.0000
Supplier:  Merck
CAS Number:  
Health
Trypan blue for microscopy, Sigma-Aldrich®
Catalog # 1.11732.0000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Dye content (spectrophotometrically):
    ≥ 80 %
  • Identity sodium:
    passes test
  • Identity UV/VIS-Spectrum:
    passes test
  • Absorption maximum λ max (water):
    590 - 600 nm
  • Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water):
    650 - 820
  • TLC-Test:
    passes test
  • Loss on drying (110 °C):
    ≤ 15 %
  • Suitability for microscopy:
    passes test

Specification Test Results

About this item

 :
 :960.82 g/mol
 :M-Clarity™ quality level = MQ300
EINECS:200-786-7
 :GHS08
Storage Temperature:Ambient
 :1
CAS Number:72-57-1
Formula:C₃₄H₂₄N₆Na₄O₁₄S₄
MDL Number:MFCD00003969