Order Entry
Italy
ContactUsLinkComponent
Acetophenone-p-toluenesulphonylhydrazone ≥99.0% (by titrimetric analysis)
Acetophenone-p-toluenesulphonylhydrazone ≥99.0% (by titrimetric analysis)
Catalog # TCIAA2174-5G
Supplier:  TCI
CAS Number:  
undefined
Acetophenone-p-toluenesulphonylhydrazone ≥99.0% (by titrimetric analysis)
Catalog # TCIAA2174-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 99.0 %

Specification Test Results

About this item

 :
 :288.36 g/mol
CAS Number:4545-21-5
Formula:C₁₅H₁₆N₂O₂S
MDL Number:MFCD01140191