Inserisci ordine
Italy
ContactUsLinkComponent
2,2'-Dihydroxy-4-methoxybenzophenone ≥98.0% (by GC)
Irritant
Health
2,2'-Dihydroxy-4-methoxybenzophenone ≥98.0% (by GC)
Catalog # TCIAD0586-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
2,2'-Dihydroxy-4-methoxybenzophenone ≥98.0% (by GC)
Catalog # TCIAD0586-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    71.0 to 75.0 deg-C
  • Solubility in Toluene:
    almost transparency

Specification Test Results

About this item

 :
 :244.246 g/mol
Merck Index:14,03303
Boiling Pt:175°C
EINECS:205-026-8
 :GHS07
 :GHS08
CAS Number:131-53-3
Formula:C₁₄H₁₂O₄
MDL Number:MFCD00002218