Order Entry
Italy
ContactUsLinkComponent
3-Butynyl toluene-4-sulphonate ≥97.0% (by GC)
Irritant
3-Butynyl toluene-4-sulphonate ≥97.0% (by GC)
Catalog # TCIAT2046-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-Butynyl toluene-4-sulphonate ≥97.0% (by GC)
Catalog # TCIAT2046-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :224.28 g/mol
 :GHS07
CAS Number:23418-85-1
Formula:C₁₁H₁₂O₃S
MDL Number:MFCD00041687