3-Butynyl toluene-4-sulphonate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23418-85-1
:
Formula:C₁₁H₁₂O₃S
MDL Number:MFCD00041687
:224.28 g/mol
Boiling Pt:128 °C (0.01 mmHg)
Density:1.012
Flash Pt:>100 °C (212 °F)
Storage Temperature:Ambient
Related Information
Frequently Bought Together