2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:106797-53-9
:
Formula:C₁₂H₁₆O₄
MDL Number:MFCD00085267
:224.2530 g/mol
EINECS:402-670-3
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:89.0 to 93.0 deg-C
Specification Test Results
Frequently Bought Together