

2-Amino-3'-nitroacetophenone hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36765-84-1
:
Formula:C₈H₉ClN₂O₃
MDL Number:MFCD00051997
Melting Pt:210…212 °C (dec.)
ADR:8,III
:216.62 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together