
4,4'-Dimethoxydiphenylamine ≥98.0% (by GC, titration analysis)
: TCI
:101-70-2
:
:C₁₄H₁₅NO₂
:MFCD00014895
:229.28 g/mol
:202-968-1
:GHS07
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:100.0 to 104.0 deg-C
Frequently Bought Together