

4-Iodo-1-methyl-1H-pyrazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39806-90-1
:
Formula:C₄H₅IN₂
MDL Number:MFCD05663860
:208.0000 g/mol
Melting Pt:61…67 °C
:GHS07
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white Semitransparent crystals or crystalline needles
- Infrared spectrum:Conforms
- Melting point:61 to 67 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together