1-Hydroxyphenazine ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:528-71-2
:
Formula:C₁₂H₈N₂O
MDL Number:MFCD00059692
Merck Index:14,04645
Specification Test Results
- Appearance:Light yellow to Brown powder to crystaline
- Purity (GC):min. 95.0 %
- Melting point:156.0 to 159.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together