6-O-Palmitoyl-L-ascorbic acid ≥97.0% (by titrimetric analysis)
TCIAA0540-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:White to Orange to Green powder to crystal
- Purity (Iodometric Titration):min. 97.0 %
- Melting point:113.0 to 117.0 deg-C
- Specific rotation [a]20/D:+20.0 to +25.0 deg (C=1, EtOH)
- Solubility in EtOH:almost transparency
:
:414.54 g/mol
:? 250 °C
:107…117 °C
:205-305-4
:137-66-6
:C₂₂H₃₈O₇
:MFCD00005377