
2,4-Heptadecadiynoic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0667-100MG
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Amber to Dark green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:57.0 to 61.0 deg-C
Specification Test Results
About this item
:
:GHS07
Storage Temperature:Freezer
CAS Number:64034-02-2
Formula:C₁₇H₂₆O₂
MDL Number:MFCD00060064