Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Biological Buffers
Buffer, HEPES solution (1 mol/L)
Buffer, HEPES solution (1 mol/L)
Buffer, HEPES solution (1 mol/L)
Catalog # BLGI03-025-1B
Supplier:
BIOLOGICAL INDUSTRIES
CAS Number:
Expand image
Buffer, HEPES solution (1 mol/L)
Catalog # BLGI03-025-1B
Supplier:
BIOLOGICAL INDUSTRIES
CAS Number:
Specifications
Pk:
1
Description:
HEPES buffer solution (1 mol/l)
Specifications
About this item
Product Details
Product Details