D-(+)-Glucurono-6,3-lactone ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:32449-92-6
:
Formula:C₆H₈O₆
MDL Number:MFCD00135622
:176.12 g/mol
Merck Index:14,04467
EINECS:251-053-3
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization back titration):min. 98.0 %
- Specific rotation [a]20/D:+17.5 to +19.5 deg (C=5, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together