



(Trimethylsilyl)diazomethane ∼10% (w/w) ∼0.6 mol/L in hexane
: TCI
:18107-18-1
:
:C₄H₁₀N₂Si
:MFCD00053946
:3,II
:114.22 g/mol
:2924
:GHS02
:GHS05
:GHS07
:GHS08
:Refrigerator
- Appearance:Light yellow to Yellow clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
- High boiling impurities:to pass test
Frequently Bought Together