

4-(1-Pyrrolidinyl)pyridine 98%
Catalog # ACRO200010050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to yellow to light brown Crystals or crystalline mass and/or chunks
- Infrared spectrum:Conforms
- Melting point:54 to 58 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:148.21 g/mol
Boiling Pt:171…173 °C (27mbar)
Melting Pt:54…58 °C
Flash Pt:112 °C
Storage Temperature:Refrigerator
EINECS:219-534-2
UN:2923
ADR:8,II
:GHS06
:GHS05
CAS Number:2456-81-7
Formula:C₉H₁₂N₂
MDL Number:MFCD00006415