Order Entry
Italy
ContactUsLinkComponent
Boron trifluoride ethylamine complex ≥97.0% (by total nitrogen basis)
Corrosive
Toxic
Boron trifluoride ethylamine complex ≥97.0% (by total nitrogen basis)
Catalog # TCIAE0399-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
Boron trifluoride ethylamine complex ≥97.0% (by total nitrogen basis)
Catalog # TCIAE0399-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to lump
  • Purity (with Total Nitrogen):
    min. 97.0 %

Specification Test Results

About this item

 :
Boiling Pt:148°C
EINECS:200-852-5
UN:2923
ADR:8,II
 :GHS05
 :GHS06
CAS Number:75-23-0
Formula:C₂H₇BF₃N
MDL Number:MFCD00144277