Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Diisopropyl tartrate
L(+)-Diisopropyl tartrate ≥98%
L(+)-Diisopropyl tartrate ≥98%
:
Thermo Fisher Scientific
L(+)-Diisopropyl tartrate ≥98%
:
Thermo Fisher Scientific
:
2217-15-4
:
:
C₁₀H₁₈O₆
:
MFCD00064450
:
234.25 g/mol
:
150…151 °C (11 mmHg)
:
1.117
:
110 °C (230 °F)
:
Ambient
:
218-709-0
msds_ALFAA16941
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Methanol ≥99.9% (by GC), LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Petroleum spirit, 40…60 °C, AnalaR NORMAPUR® ACS analytical reagent (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Acetonitrile, LiChrosolv®, Isocratic Grade for liquid chromatography, Supelco®
: Merck
Toluene, EMPLURA®, extra pure, Supelco®
: Merck
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
2,2,4-Trimethylpentane, SupraSolv® for GC-ECD/FID, Supelco®
: Merck
Dimethyl sulphoxide, SupraSolv® for headspace gas chromatography, Supelco®
: Merck
Methanol, Prepsolv® for preparative chromatography, Supelco®
: Merck
Acetonitrile ≥99%, TECHNICAL
: VWR Chemicals