
Di-tert-butyl iminodicarboxylate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:51779-32-9
:
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD00043309
:217.27 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
Specification Test Results
Related Information
Frequently Bought Together