
1,4-Dicyano-2-butene ≥98.0% (by GC)
: TCI
:1119-85-3
:
:C₆H₆N₂
:MFCD00001964
:106.1252 g/mol
:120°C
:214-287-7
:3439
:6.1,III
:GHS06
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:76.0 to 79.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together