Affinity chromatography columns, HiTrap™ Benzamidine FF
Supplier: Cytiva
Certificates
About this item
HiTrap Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
- Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates
- High binding capacity
- It can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC system
HiTrap Benzamidine FF (high sub) are prepacked 1 ml and 5 ml columns for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.
The medium, Benzamidine Sepharose 4 Fast Flow (high sub), is also available as lab packages and is an excellent choice for scale-up.
Specifications
- Catalog No:
- 17-5143-01
- 17-5143-02
- 17-5144-01
- Max. pressure:
- 5 bar
- 5 bar
- 5 bar
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- Storage:
- 2 to 8 °C, 20% Ethanol, 0,05 M Sodium Acetate, pH 4,0
- 2 to 8 °C, 20% Ethanol, 0,05 M Sodium Acetate, pH 4,0
- 2 to 8 °C, 20% Ethanol, 0,05 M Sodium Acetate, pH 4,0
Specifications
Related Information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together