Rapid-hyb buffer
Rapid-hyb Buffer sufficient for 4000 cm² membrane, 500 mL
RPN1636
: Cytiva
A major limitation of membrane hybridisation is speed. At commonly used probe concentrations (2 to 10 ng/ml), hybridisation normally takes 16 hours to detect single-copy gene sequences in mammalian DNA. Use of Rapid-hyb buffer reduces this time to 2 hours using similar probe concentrations.
Store at ambient temperature. Under these conditions the product is stable for at least 3 months.
- Pk:500 mL
- Description:Rapid-hyb Buffer sufficient for 4000 cm² membrane
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..