Order Entry
Italy
ContactUsLinkComponent
Flame-retardant fleece lining for jackets, Royston 7220
Flame-retardant fleece lining for jackets, Royston 7220
Supplier:  SIOEN
Certificates
undefined
Flame-retardant fleece lining for jackets, Royston 7220
Supplier:  SIOEN
Specifications
  • Catalog No:
  • 113-7218
  • 113-7217
  • 113-7220
  • 113-7222
  • 113-7219
  • 113-7221
  • Model:
  • Royston 7220
  • Royston 7220
  • Royston 7220
  • Royston 7220
  • Royston 7220
  • Royston 7220
  • Garment application:
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Fire protection
  • Risk area:
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Flame-retardant
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Weight:
  • 285 g/m²
  • 285 g/m²
  • 285 g/m²
  • 285 g/m²
  • 285 g/m²
  • 285 g/m²