Order Entry
Italy
ContactUsLinkComponent
VWR® Contact Plate Weight Applicator, Contact Weight
VWR® Contact Plate Weight Applicator, Contact Weight
Catalog # 710-0961
Supplier:  VWR Collection
CAS Number:  
undefined
VWR® Contact Plate Weight Applicator, Contact Weight
Catalog # 710-0961
Supplier:  VWR Collection
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Contact plate applicator, Contact Weight
Specifications
About this item
Used in combination with contact plates, the contact weight enables the standardisation of microbiological surface control between operators. It applies the same constant pressure for a pre-set time on a contact plate. This ensures results are directly comparable. The contact weight meets the ISO 18593 standard, and has a sterilisable stainless steel base and pull-out digital timer.
  • Dry heat or steam sterilisable stainless steel base, with pull-out digital timer
  • Can be used with different types of contact plates
  • According to ISO 18593
  • Weight: 500 g
Timer included provided without the certificate of calibration; if a calibration is required it is necessary to buy the article 609-0184 as accessory that includes the calibration.