Order Entry
Italy
ContactUsLinkComponent
Anion exchange chromatography column, HiScreen™ Capto™ Q ImpRes
Anion exchange chromatography column, HiScreen™ Capto™ Q ImpRes
Catalog # 17-5470-15
Supplier:  Cytiva
undefined
Anion exchange chromatography column, HiScreen™ Capto™ Q ImpRes
Catalog # 17-5470-15
Supplier:  Cytiva

Specifications

  • Max. pressure:
    4 bar
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ Capto™ Q ImpRes
  • Storage:
    4 to 30 °C, 20% Ethanol
  • Bed dimensions:
    7,7×100 mm
  • Flow velocity:
    100 - 300 cm/h
  • Pk:
    1

Specifications

About this item

HiScreen™ Capto™ Q ImpRes columns are prepacked with Capto™ Q ImpRes strong anion exchange BioProcess medium. An excellent choice for method optimization and parameter screening.

  • Capto™ Q ImpRes is designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification
  • Columns with bed height of 10 cm, is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity