VWR® BANDit™ Protein Stain Blue
Catalog # K217-1L
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pk:1 L
- Description:Protein stain for 50 mini gels
- Seq:002
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
About this item
Blue BANDit is a safe and environmentally friendly alternative to traditional Coomassie® Blue staining.
- Reduces handling of hazardous chemicals: No methanol or acetic acid required
- High sensitivity, quick, simple procedure