Order Entry
Italy
ContactUsLinkComponent
2',3'-Dideoxyadenosine ≥98%
2',3'-Dideoxyadenosine ≥98%
Catalog # J64573.MB
CAS Number:  
2',3'-Dideoxyadenosine ≥98%
Catalog # J64573.MB
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg

Specifications

About this item

 :
Merck Index:14,3101
Melting Pt:181…184 °C
 :235.24 g/mol
EINECS:223-853-2
CAS Number:4097-22-7
Formula:C₁₀H₁₃N₅O₂
MDL Number:MFCD00010534