Order Entry
Italy
ContactUsLinkComponent
 
Irritant
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%
Codice catalogo J63427.MC
CAS Number:  
Irritant
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%
Codice catalogo J63427.MC
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :550.09 g/mol
EINECS:243-347-5
 :GHS07
Storage Temperature:Freezer
CAS Number:19817-92-6
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00044310