Order Entry
Italy
ContactUsLinkComponent
Triflumuron ≥98%
Triflumuron ≥98%
Catalog # J63112.22
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  
Triflumuron ≥98%
Catalog # J63112.22
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifiche

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifiche

Informazioni su questo articolo

Numero CAS:64628-44-0
 :
Formula:C₁₅H₁₀ClF₃N₂O₃
 :358.7 g/mol
IndexMerck:14,9678
Punto di fusione:198 °C
temperatura di conservazione:Ambient
EINECS:264-980-3
Numero MDL:MFCD00072496